Learn More
Aviva Systems Biology Corp EIF2S2 Antibody - N-terminal region (ARP40502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2S2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF2S2. Alternate names: DKFZp686L18198, EIF2, EIF2B, EIF2beta, MGC8508, PPP1R67ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53205