Learn More
Aviva Systems Biology Corp EIF2S3 Antibody - N-terminal region (ARP46056_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46056_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2S3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF2S3. Alternate names: EIF2, EIF2G, EIF2gamma, eIF-2gAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59200