Learn More
Aviva Systems Biology Corp Eif3d Antibody - N-terminal region (ARP62385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eif3d. It was validated on Western Blot. . Alternate names: Eif3s7, MGC93918ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-666