Learn More
Aviva Systems Biology Corp EIF3E Antibody - middle region (ARP48176_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48176_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EIF3E. Alternate names: EIF3-P48, EIF3S6, INT6, eIF3-p46ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53543