Learn More
Aviva Systems Biology Corp EIF3F Antibody - C-terminal region (ARP61971_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61971_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3F. It was validated on Western Blot. . Alternate names: EIF3S5, eIF3-p47ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-526