Learn More
Aviva Systems Biology Corp EIF3G Antibody - middle region (ARP40485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3G. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EIF3G. Alternate names: EIF3-P42, EIF3S4, eIF3-delta, eIF3-p44ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57479