Learn More
Aviva Systems Biology Corp EIF3J Antibody - C-terminal region (ARP65317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3J. It was validated on Western Blot. . Alternate names: EIF3S1, eIF3-alpha, eIF3-p35ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-470