Learn More
Aviva Systems Biology Corp EIF3K Antibody - C-terminal region (ARP62169_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62169_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3K. It was validated on Western Blot. . Alternate names: ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-596