Learn More
Aviva Systems Biology Corp EIF3M Antibody - N-terminal region (ARP48293_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48293_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3M. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EIF3M. Alternate names: B5, FLJ29030, GA17, PCID1, hfl-B5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53548