Learn More
Aviva Systems Biology Corp EIF3S9 Antibody - C-terminal region (ARP40483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF3S9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human EIF3S9. Alternate names: PRT1, EIF3S9, EIF3-ETA, EIF3-P110, EIF3-P116ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53203