Learn More
Aviva Systems Biology Corp EIF4A1 Antibody - middle region (ARP48065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EIF4A1. Alternate names: DDX2A, EIF-4A, EIF4A, eIF4A-I, eIF-4A-IApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-285-344