Learn More
Aviva Systems Biology Corp EIF4A2 Antibody - N-terminal region (ARP40376_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF4A2. Alternate names: DDX2B, EIF4A, EIF4F, BM-010, eIF4A-II, eIF-4A-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57418