Learn More
Aviva Systems Biology Corp EIF4A3 Antibody - N-terminal region (ARP36421_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36421_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4A3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EIF4A3. Alternate names: DDX48, DKFZp686O16189, KIAA0111, MGC10862, NMP265, NUK34, eIF4AIIIApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-284-497