Learn More
Medchemexpress LLC eIF4A3-IN-1 | 2095486-67-0 | 100.0% | 588.88 g·mol⁻¹ | C29H23BrClN5O2 | 10 MG
SDP
Catalog No. 5000246532
Encompass_Preferred
eIF4A3-IN-1 is a small-molecule inhibitor of eukaryotic initiation factor 4A-3 (eIF4A3). It binds a non-ATP site on eIF4A3, inhibits nonsense-mediated RNA decay in cellular assays, and has reported antitumor and analgesic activity. The compound is supplied as a white to off-white solid or as a DMSO solution, with high purity and recommended low-temperature storage for long-term stability.
  • Selective eIF4A3 inhibitor that binds a non-ATP binding site.
  • Inhibits nonsense-mediated RNA decay in cellular assays.
  • Exhibits antitumor and analgesic activity in preclinical studies.
  • High purity suitable for research applications.
  • Available as solid or DMSO solution for convenient use.
  • Recommended low-temperature storage to maintain stability.
Catalog No. 50-002-46532 Supplier Medchemexpress LLC Supplier No. HY10151310MG
eIF4A3-IN-1 is a small-molecule inhibitor of eukaryotic initiation factor 4A-3 (eIF4A3). It binds a non-ATP site on eIF4A3, inhibits nonsense-mediated RNA decay in cellular assays, and has reported antitumor and analgesic activity. The compound is supplied as a white to off-white solid or as a DMSO solution, with high purity and recommended low-temperature storage for long-term stability.
  • Selective eIF4A3 inhibitor that binds a non-ATP binding site.
  • Inhibits nonsense-mediated RNA decay in cellular assays.
  • Exhibits antitumor and analgesic activity in preclinical studies.
  • High purity suitable for research applications.
  • Available as solid or DMSO solution for convenient use.
  • Recommended low-temperature storage to maintain stability.