Learn More
Medchemexpress LLC HY-101513 5mg Medchemexpress, eIF4A3-IN-1 CAS:2095486-67-0 Purity:>98%
SDP
Catalog No. 502029497
Encompass_Preferred
Medchemexpress, HY-101513 5mg eIF4A3-IN-1 CAS:2095486-67-0 eIF4A3-IN-1 (compound 53a) is a selective eukaryotic initiation factor 4A3 (eIF4A3) inhibitor (IC50=0.26 μM; Kd=0.043 μM), which binds to a non-ATP binding site of eIF4A3 and shows significant cellular nonsense-mediated RNA decay (NMD) inhibition at 10 and 3 μM and can be as a probe for further study of eIF4A3, the exon junction complex (EJC), and NMD. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9497 Supplier Medchemexpress LLC Supplier No. HY1015135MG
Medchemexpress, HY-101513 5mg eIF4A3-IN-1 CAS:2095486-67-0 eIF4A3-IN-1 (compound 53a) is a selective eukaryotic initiation factor 4A3 (eIF4A3) inhibitor (IC50=0.26 μM; Kd=0.043 μM), which binds to a non-ATP binding site of eIF4A3 and shows significant cellular nonsense-mediated RNA decay (NMD) inhibition at 10 and 3 μM and can be as a probe for further study of eIF4A3, the exon junction complex (EJC), and NMD. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.