Learn More
Medchemexpress LLC HY-101785 100mg Medchemexpress, eIF4A3-IN-2 CAS:2095677-20-4 Purity:>98%
SDP
Catalog No. 501932215
Encompass_Preferred
Medchemexpress, HY-101785 100mg eIF4A3-IN-2 CAS:2095677-20-4 eIF4A3-IN-2 is a highly selective and noncompetitive eukaryotic initiation factor 4A-3 (eIF4A3) inhibitor with an IC 50 of 110 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2215 Supplier Medchemexpress LLC Supplier No. HY101785100MG
Medchemexpress, HY-101785 100mg eIF4A3-IN-2 CAS:2095677-20-4 eIF4A3-IN-2 is a highly selective and noncompetitive eukaryotic initiation factor 4A-3 (eIF4A3) inhibitor with an IC 50 of 110 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.