Learn More
Medchemexpress LLC eIF4A3-IN-8 | 18474-59-4 | 99.9% | 175.18 | 100 MG
SDP
Catalog No. 5000404204
Encompass_Preferred
eIF4A3-IN-8 is a selective ATP-competitive eukaryotic initiation factor 4A3 (eIF4A3) inhibitor. It can serve as a valuable chemical probe to elucidate the detailed function of eIF4A3 and the exon junction complex (EJC).
  • Selective ATP-competitive inhibitor
  • Chemical probe for elucidating eIF4A3 and EJC function
Catalog No. 50-004-04204 Supplier Medchemexpress LLC Supplier No. HY33838100MG
eIF4A3-IN-8 is a selective ATP-competitive eukaryotic initiation factor 4A3 (eIF4A3) inhibitor. It can serve as a valuable chemical probe to elucidate the detailed function of eIF4A3 and the exon junction complex (EJC).
  • Selective ATP-competitive inhibitor
  • Chemical probe for elucidating eIF4A3 and EJC function