Learn More
Medchemexpress LLC eIF4A3-IN-8 | 18474-59-4 | 99.9% | 175.18 | 250 MG
SDP
Catalog No. 5000402318
Encompass_Preferred
eIF4A3-IN-8 is a selective ATP-competitive eukaryotic initiation factor 4A3 (eIF4A3) inhibitor. It can serve as a valuable chemical probe to elucidate the detailed function of eIF4A3 and EJC (exon junction complex).
  • Selective ATP-competitive eIF4A3 inhibitor
  • Can be used as a chemical probe to study eIF4A3 and EJC
Catalog No. 50-004-02318 Supplier Medchemexpress LLC Supplier No. 250MGHY33838
eIF4A3-IN-8 is a selective ATP-competitive eukaryotic initiation factor 4A3 (eIF4A3) inhibitor. It can serve as a valuable chemical probe to elucidate the detailed function of eIF4A3 and EJC (exon junction complex).
  • Selective ATP-competitive eIF4A3 inhibitor
  • Can be used as a chemical probe to study eIF4A3 and EJC