Learn More
Enzo Life Sciences eIF4E BP 2 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPMA112E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human eIF4E-BP2. The sequence is completely conserved in mouse. UniProt ID: Q13542. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Dog, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (4µg/ml colorimetric, 4µg/ml ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In borate buffered saline, pH 8.4, containing 25mM sodium borate, 100mM boric acid, 75mM sodium chloride, and 5mM EDTA. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7282