Learn More
Aviva Systems Biology Corp EIF4E2 Antibody - N-terminal region (ARP40555_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40555_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4E2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EIF4E2. Alternate names: 4EHP, IF4e, 4E-LP, EIF4EL3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57521