Learn More
Aviva Systems Biology Corp EIF4E2 Antibody - N-terminal region (ARP58898_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58898_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4E2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EIF4E2. Alternate names: 4E-LP, 4EHP, EIF4EL3, IF4eApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-520