Learn More
Aviva Systems Biology Corp Eif4ebp1 Antibody - N-terminal region (ARP52033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eif4ebp1. It was validated on Western Blot. . Alternate names: 4e-bp1, AA959816, PHAS-IApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-627