Learn More
Aviva Systems Biology Corp EIF4ENIF1 Antibody - N-terminal region (ARP57349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4ENIF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF4ENIF1. Alternate names: 4E-T, Clast4, FLJ21601, FLJ26551ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53960