Learn More
ABclonal Technology EIF4ENIF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A151751000UL
Encompass_Preferred
The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic, its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07040