Learn More
Aviva Systems Biology Corp EIF4G1 Antibody - middle region (ARP40562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4G1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EIF4G1. Alternate names: DKFZp686A1451, EIF4F, EIF4G, p220, P220, EIF4GI, PARK18, EIF-4G1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57524