Learn More
Aviva Systems Biology Corp EIF4G2 Antibody - N-terminal region (ARP40362_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40362_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4G2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF4G2. Alternate names: AAG1, DAP5, FLJ41344, NAT1, p97, P97ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57411