Learn More
Aviva Systems Biology Corp EIF4H Antibody - C-terminal region (ARP40977_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40977_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF4H. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human EIF4H. Alternate names: KIAA0038, WBSCR1, WSCR1, eIF-4HApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57683