Learn More
Aviva Systems Biology Corp EIF5A2 Antibody - N-terminal region (ARP57402_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57402_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF5A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF5A2. Alternate names: EIF-5A2, eIF5AIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53959