Learn More
Aviva Systems Biology Corp EIF6 Antibody - C-terminal region (ARP61387_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61387_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF6. It was validated on Western Blot. . Alternate names: 2, CAB, EIF3A, ITGB4BP, b, b(2)gcn, gcn, p27BBP, eIF-6, p27(BBP)ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-287-360