Learn More
BRAIN RESEARCH LABORATORIES EIGHT-SECTION STAINING NETS
Catalog No. NC2383527
Encompass
Eight-section Staining Nets3½″ dia. x 1½″ high with 32mesh nettingStaining Nets made exclusively by Brain Research Laboratories. Constructed of a clear, bonded plexiglass, permitting easy see-thru viewing of tissue.Recommended cleaning: Mild detergents only. No harsh chemicals or acids.CAUTION: Staining Nets are not suitable for xylenes or heat, not dishwasher safe.All Staining Nets are made to order and require 2-3 days before shipping.Made in the USA32mesh netting – 32 holes per inch / 64mesh netting – 64 holes per inch
Catalog No. NC2383527 Supplier BRAIN RESEARCH LABORATORIES Supplier No. 8094
Add to Cart
Edge
Add to Cart
Eight-section Staining Nets3½″ dia. x 1½″ high with 32mesh nettingStaining Nets made exclusively by Brain Research Laboratories. Constructed of a clear, bonded plexiglass, permitting easy see-thru viewing of tissue.Recommended cleaning: Mild detergents only. No harsh chemicals or acids.CAUTION: Staining Nets are not suitable for xylenes or heat, not dishwasher safe.All Staining Nets are made to order and require 2-3 days before shipping.Made in the USA32mesh netting – 32 holes per inch / 64mesh netting – 64 holes per inch