Learn More
Andwin Scientific EIGHT TUBE DECAPPER
SDP
Catalog No. 50819902
Encompass
The manual Eight tube decapper help users open tubes rapidly with no difficulty. This product can be used with Micronic TPE Push Caps, TPE LyoCaps and EVA Push Caps in 96-well format for 0.50ml, 0.75ml, 1.10ml, 1.40ml, 2.00ml, and 2.50ml tubes. It can also be used to decap TPE Push Caps made by other brands.
Catalog No. 50-819-902 Supplier Andwin Scientific Supplier No. 17654001
The manual Eight tube decapper help users open tubes rapidly with no difficulty. This product can be used with Micronic TPE Push Caps, TPE LyoCaps and EVA Push Caps in 96-well format for 0.50ml, 0.75ml, 1.10ml, 1.40ml, 2.00ml, and 2.50ml tubes. It can also be used to decap TPE Push Caps made by other brands.