Learn More
ATCC Klebsiella pneumoniae
Supplier:  ATCC BAA1144
Encompass
Klebsiella pneumoniae strain UMJMH14 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This strain produces a low level of AmpC ?-lactamase and it can be used as a control isolate for the AmpC disk test.
Catalog No. 50-238-0142