Learn More
Bulbman Inc Eiko 1460 Bulb 6.5V 2.75A S8 Incandescent Bulb
SDP
Catalog No. NC1210547
Encompass_Preferred
This Eiko 40279 is a ANSI code 1460 bulb. It uses 18 watts and 6.5 volts. It has a P30D base, and an average rated life of 100 hours. It has a S8 lamp shape, and is an incandescent bulb.
Catalog No. NC1210547 Supplier Bulbman Inc Supplier No. 107171
This Eiko 40279 is a ANSI code 1460 bulb. It uses 18 watts and 6.5 volts. It has a P30D base, and an average rated life of 100 hours. It has a S8 lamp shape, and is an incandescent bulb.