Learn More
Bulbman Inc Sylvania 6 Volt, 15 Watt 8017 Bulb, Ba15D Base
SDP
Catalog No. NC9431329
Encompass_Preferred
Bulb; 6V; 15W; BA15D Base; T6 Shape; 1000 Life hr.
Catalog No. NC9431329 Supplier Bulbman Inc Supplier No. 102034
Bulb; 6V; 15W; BA15D Base; T6 Shape; 1000 Life hr.