Learn More
Medchemexpress LLC HY-101840 10mg Medchemexpress, EIPA CAS:1154-25-2 Purity:>98%
SDP
Catalog No. 501967882
Encompass_Preferred
Medchemexpress, HY-101840 10mg EIPA CAS:1154-25-2 EIPA (L593754; MH 12-43) is a TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA (L593754; MH 12-43) also inhibits Na+/H+-exchanger (NHE) and macropinocytosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7882 Supplier Medchemexpress LLC Supplier No. HY10184010MG
Medchemexpress, HY-101840 10mg EIPA CAS:1154-25-2 EIPA (L593754; MH 12-43) is a TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA (L593754; MH 12-43) also inhibits Na+/H+-exchanger (NHE) and macropinocytosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.