Learn More
Medchemexpress LLC HY-101840A 10mg Medchemexpress, EIPA (hydrochloride) CAS:1345839-28-2 Purity:>98%
SDP
Catalog No. 501967883
Encompass_Preferred
Medchemexpress, HY-101840A 10mg EIPA (HCl) CAS:1345839-28-2 EIPA hydrochloride (L593754 hydrochloride) is a TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA also inhibits Na+/H+-exchanger (NHE) and macropinocytosis[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7883 Supplier Medchemexpress LLC Supplier No. HY101840A10MG
Medchemexpress, HY-101840A 10mg EIPA (HCl) CAS:1345839-28-2 EIPA hydrochloride (L593754 hydrochloride) is a TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA also inhibits Na+/H+-exchanger (NHE) and macropinocytosis[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.