Learn More
Medchemexpress LLC HY-101840A 5mg Medchemexpress, EIPA (hydrochloride) CAS:1345839-28-2 Purity:>98%
SDP
Catalog No. 501932224
Encompass_Preferred
Medchemexpress, HY-101840A 5mg EIPA (hydrochloride) CAS:1345839-28-2 EIPA hydrochloride (L593754 hydrochloride) is a TRPP3 channel inhibitor with an IC 50 of 10.5 μM [1] . EIPA also inhibits Na + /H + -exchanger ( NHE ) [2] and macropinocytosis [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2224 Supplier Medchemexpress LLC Supplier No. HY101840A5MG
Medchemexpress, HY-101840A 5mg EIPA (hydrochloride) CAS:1345839-28-2 EIPA hydrochloride (L593754 hydrochloride) is a TRPP3 channel inhibitor with an IC 50 of 10.5 μM [1] . EIPA also inhibits Na + /H + -exchanger ( NHE ) [2] and macropinocytosis [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.