Learn More
Medchemexpress LLC EIPA hydrochloride | 1345839-28-2 | 98.3% | 336.22 | C11H19Cl2N7O | 1 ML
SDP
Catalog No. 5000248902
Encompass_Preferred
EIPA hydrochloride (CAS 1345839-28-2) is a small-molecule research reagent used in cell biology to inhibit macropinocytosis and modulate ion channel activity. It is used in studies of inflammation, autophagy, and cancer research. The compound has molecular formula C11H19Cl2N7O and molecular weight 336.22 g/mol.
  • High reported purity: 98.3% (HPLC where stated).
  • Available as concentrated stock solution formats for laboratory use.
  • Documented chemical identifiers for cross-referencing (CAS and InChIKey).
  • Useful in studies of macropinocytosis, autophagy, and ion channel modulation.
  • Datasheet provides preparation guidance for common stock concentrations.
Catalog No. 50-002-48902 Supplier Medchemexpress LLC Supplier No. HY101840A10MM 1ML
EIPA hydrochloride (CAS 1345839-28-2) is a small-molecule research reagent used in cell biology to inhibit macropinocytosis and modulate ion channel activity. It is used in studies of inflammation, autophagy, and cancer research. The compound has molecular formula C11H19Cl2N7O and molecular weight 336.22 g/mol.
  • High reported purity: 98.3% (HPLC where stated).
  • Available as concentrated stock solution formats for laboratory use.
  • Documented chemical identifiers for cross-referencing (CAS and InChIKey).
  • Useful in studies of macropinocytosis, autophagy, and ion channel modulation.
  • Datasheet provides preparation guidance for common stock concentrations.