Learn More
Bulbman Inc EJL 24200W HALOGEN LAMP
SDP
Catalog No. NC1299169
Encompass_Preferred
EJL 24 volt, 200 watt, with a GX5.3 base optical reflector halogen lamp
Catalog No. NC1299169 Supplier Bulbman Inc Supplier No. 103378
EJL 24 volt, 200 watt, with a GX5.3 base optical reflector halogen lamp