Learn More
Electron Microscopy Sciences Kodak 3 Color Ribbon
SDP
Supplier:  Electron Microscopy Sciences 89050
Encompass
Media product for outstanding image stability and near-photographic quality continuous tone thermal prints. Feature excellent color reproduction, including superior flesh tones and durability. The unique Kodak XtraLife Ribbon overcoats prints with a rugged transparent layer which prevents destructive dye transfer as well as it resists fingerprints and fading.
Catalog No. 50-267-76