Learn More
Biomatik Corporation Mouse Slit Homolog 3 (Slit3) ELISA Kit
Supplier:  Biomatik Corporation EKU11431
Encompass_Preferred
Species Reactivity: Mouse, Format: 96-Well Plate, Assay Type: Double-antibody Sandwich, UniProt ID: Q9WVB4, Restriction: For Research Use Only. Not for use in diagnostic procedures. Not for human or animal drug or food use.
Catalog No. NC1728828