Learn More
Cole-Parmer Cole-Parmer elements Erlenmeyer Flask, Glass, 4000 mL, 1/pk
Catalog No. NC1765737 Shop All Cole-Parmer Products
Encompass_Preferred
Made with durable borosilicate 3.3 glass and featuring a heavy-duty rim with narrow-mouth opening there are very few applications these flasks cannot handle.
Catalog No. NC1765737 Supplier Cole-Parmer Supplier No. 3450265
Made with durable borosilicate 3.3 glass and featuring a heavy-duty rim with narrow-mouth opening there are very few applications these flasks cannot handle.