Learn More
Cole-Parmer Essentials Erlenmeyer Flask, Glass, 5000 mL; 1/PK
Supplier:  Cole-Parmer 3450266
Encompass_Preferred
Made with durable borosilicate 3.3 glass and featuring a heavy-duty rim with narrow-mouth opening there are very few applications these flasks cannot handle.
Catalog No. NC1879859