Learn More
ATCC EL4; Lymphoma; Mouse
Supplier:  ATCC TIB39
Encompass
EL4 is a T lymphoblast that was established from a lymphoma induced in a C57BL mouse by 9,10-dimethyl-1,2-benzanthracene. This cell line can be used in immunology research.
Catalog No. 50-238-4209