Learn More
Medchemexpress LLC Ela-32(human) TFA | 99.9% | 3967.82 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP2196A5MG
Encompass_Preferred
ELA-32(human) TFA is a potent, high affinity apelin receptor agonist. It has an IC50 of 0.27 nM and a Kd of 0.51 nM, and does not bind to GPR15 and GPR25.
  • Activates the PI3K/AKT pathway
  • Promotes self-renewal of hESCs through cell-cycle progression and protein translation
  • Potentiates the TGFβ pathway, priming hESCs toward the endoderm lineage
  • Stimulates angiogenesis in HUVEC cells
Catalog No. 50-004-22719