Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alanine Aminotransferase (ALT/GPT) Activity Kit

Catalog No. EEA001
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA001 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA001 Supplier Invitrogen™ Supplier No. EEA001
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Alanine Aminotransferase (ALT/GPT) Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ALT activity in animal serum, plasma, tissue, cultured cells, and cell culture supernatant. ALT is an important pyridoxal phosphate dependent enzyme in the intermediate metabolism of glucose and proteins. Clinically, the activity of serum alanine aminotransferase is often used as a marker for alcoholic liver disease, liver cirrhosis, and acute viral hepatitis.

In this assay, ALT catalyzes the amino conversion reaction between alanine and α-ketoglutaric acid to produce pyruvic acid and glutamic acid. Phenylhydrazine is added to form phenylhydrazone with pyruvic acid. Phenylhydrazone is a reddish brown solution under alkaline conditions, and ALT activity can be calculated by measuring the OD values at 510 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.75–72.3 IU/L
Assay Sensitivity 0.75 IU/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 50 min.
Interassay CV 9
Intraassay CV 5.3
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Alanine Aminotransferase (ALT/GPT)
Total Assay Time 80 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.