Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aspartate Aminotransferase (AST/GOT) Activity Kit

Catalog No. EEA003
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA003 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA003 Supplier Invitrogen™ Supplier No. EEA003
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Aspartate Aminotransferase (AST/GOT) Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of aspartate aminotransferase activity in various sample types. AST/GOT is a key enzyme in nitrogen metabolism, which is widely found in plasma and body tissues, including liver, heart, skeletal muscle, kidney, brain, pancreas, lung, and erythrocytes. Changes in AST/GOT activity were found in acute pancreatitis, ischemic stroke, severe burns, periodontitis, acute renal disease, and motor neuron disease.

The assay is based on the ability of AST/GOT to enable alpha-ketoglutaric acid and aspartic acid to displace amino and keto groups to form glutamic acid and oxaloacetic acid. Oxaloacetic acid can decarboxylate itself to form pyroracemic acid during the reaction. Pyroracemic acid reacts with 2,4-dinitro phenyl hydrazine (DNPH) to form 2,4, dinitrophenylhydrazone, showing reddish-brown color in alkaline solution. The OD values can be measured to calculate the enzyme activity.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.1–72.3 IU/L
Assay Sensitivity 1.1 IU/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 50 min.
Interassay CV 6.8
Intraassay CV 5.3
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Aspartate Aminotransferase (AST/GOT)
Total Assay Time 80 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.