Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcium (Ca) Colorimetric Assay Kit

Catalog No. EEA006
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA006 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA006 Supplier Invitrogen™ Supplier No. EEA006
Only null left
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Calcium Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of calcium content in various sample types. Calcium is an important multifunctional signaling molecule required for biological functions such as muscle contraction, nerve conduction, and blood regulation. Calcium-mediated signal transduction is the basis of many basic processes of mammalian development and reproduction.

In this assay, calcium ions in samples bind to Methyl Thymol Blue (MTB) in an alkaline solution and form a blue complex. The blue complex has a specific absorption peak at 610 nm, and calcium content can be calculated by measuring the OD.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.07–1.2 mmol/L
Assay Sensitivity 0.07 mmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Plasma, Serum, Tissue Homogenates, Urine
For Use With (Equipment) Microplate Reader
Incubation Time 5 min.
Interassay CV 8.5
Intraassay CV 4.7
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Calcium (Ca)
Total Assay Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.