Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutathione Peroxidase (GSH-Px) Activity Kit

Catalog No. EEA010
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA010 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA010 Supplier Invitrogen™ Supplier No. EEA010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Glutathione Peroxidase (GSH-Px) Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GSH-Px activity in various sample types. GSH-Px is a crucial enzyme that plays a key role in protecting cells against oxidative damage by catalyzing the reduction of hydrogen peroxide and organic hydroperoxides using glutathione as a cofactor, protecting cell membrane structure and keeping membrane function integrity.

The activity of glutathione can be calculated by measuring the consumption of reduced glutathione. GSH can react with dinitrobenzoic acid to produce 5-thio-dinitrobenzoic acid anion, which presents with a stable yellow color. GSH can be calculated by measuring the absorbance of the anions in solution at 412 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 17.17–518.32 U/L
Assay Sensitivity 17.17 U/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Cells
For Use With (Equipment) Microplate Reader
Incubation Time 10 min.
Interassay CV 6.2
Intraassay CV 2.4
Quantity 96 tests
Sample Volume 40 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Glutathione Peroxidase (GSH-Px)
Total Assay Time 40 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.