Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Potassium (K) Turbidimetric Assay Kit

Catalog No. EEA018
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA018 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA018 Supplier Invitrogen™ Supplier No. EEA018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Potassium (K) Turbidimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of potassium content in serum, plasma, milk, tissue, cells, and other sample types. Potassium is an essential ion that plays a crucial role in various physiological processes, including muscle and nerve function, fluid balance, and cardiovascular health. Potassium deficiency and excess can each result in different signs and symptoms, including abnormal heart rhythm and various electrocardiographic abnormalities

In this assay, under alkaline conditions, potassium ions in the sample react with sodium tetraphenylborate to form potassium tetraphenylborate, a turbid white solution with little solubility. The turbidity is proportional to the potassium ion concentration, which can be calculated indirectly by measuring the OD value at 450 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.01–0.80 mmol/L
Assay Sensitivity 0.002 mmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Milk, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 5 min.
Interassay CV 6.1
Intraassay CV 1.1
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Potassium (K)
Total Assay Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.